Anti-Neurofibromin Rabbit polyclonal antibody

Supplier: Abcam
ABCAAB128054-50EA 560 EUR
ABCAAB128054-50
Anti-Neurofibromin Rabbit polyclonal antibody
Type: Primary
Antigen: Neurofibromin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR