Anti-SAMHD1 Mouse monoclonal antibody [clone: 1A1]

Supplier: Abcam
ABCAAB128107-100EA 480 EUR
ABCAAB128107-100
Anti-SAMHD1 Mouse monoclonal antibody [clone: 1A1]
Type: Primary
Antigen: SAMHD1 (SAM domain and HD domain 1)
Clonality: monoclonal
Clone: 1A1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR