Anti-HA tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Abcam
ABCAAB128131-100EA 555 EUR
ABCAAB128131-100
Anti-HA tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Primary
Antigen: HA tag (HA tag)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR