Anti-CARM1 Rabbit polyclonal antibody unconjugated

ABCAAB128851-100EA 465 EUR
ABCAAB128851-100
Anti-CARM1 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: CARM1 (coactivator-associated arginine methyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR