Anti-Prostate Secretory Protein/PSP Rabbit Monoclonal Antibody [clone: EPR7345]

Supplier: Abcam
ABCAAB128897-100EA 505 EUR
ABCAAB128897-100
Anti-Prostate Secretory Protein/PSP Rabbit Monoclonal Antibody [clone: EPR7345]
Antibodies
Type: Primary
Antigen: Prostate Secretory Protein/PSP
Clonality: monoclonal
Clone: EPR7345
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR