Anti-AQP4 Rabbit monoclonal antibody unconjugated [clone: EPR7040]

Supplier: Abcam
ABCAAB128906-100EA 575 EUR
ABCAAB128906-100
Anti-AQP4 Rabbit monoclonal antibody unconjugated [clone: EPR7040]
Antibodies
Type: Primary
Antigen: AQP4 (aquaporin 4)
Clonality: monoclonal
Clone: EPR7040
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR