Anti-ACAA2 Rabbit Monoclonal Antibody [clone: EPR6732]

Supplier: Abcam
ABCAAB128929-100EA 530 EUR
ABCAAB128929-100
Anti-ACAA2 Rabbit Monoclonal Antibody [clone: EPR6732]
Antibodies
Type: Primary
Antigen: ACAA2 (acetyl-CoA acyltransferase 2)
Clonality: monoclonal
Clone: EPR6732
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR