Anti-GSR Rabbit Monoclonal Antibody [clone: EPR7237]

Supplier: Abcam
ABCAAB128933-100EA 630 EUR
ABCAAB128933-100
Anti-GSR Rabbit Monoclonal Antibody [clone: EPR7237]
Antibodies
Type: Primary
Antigen: GSR (glutathione reductase)
Clonality: monoclonal
Clone: EPR7237
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR