Anti-SNRPA1 Rabbit Monoclonal Antibody [clone: EPR7557]

Supplier: Abcam
ABCAAB128937-100EA 515 EUR
ABCAAB128937-100
Anti-SNRPA1 Rabbit Monoclonal Antibody [clone: EPR7557]
Antibodies
Type: Primary
Antigen: SNRPA1 (small nuclear ribonucleoprotein polypeptide A')
Clonality: monoclonal
Clone: EPR7557
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR