Anti-CPS1 Rabbit Monoclonal Antibody [clone: EPR7494]

Supplier: Abcam
ABCAAB128942-100EA 515 EUR
ABCAAB128942-100
Anti-CPS1 Rabbit Monoclonal Antibody [clone: EPR7494]
Antibodies
Type: Primary
Antigen: CPS1 (carbamoyl-phosphate synthase 1, mitochondrial)
Clonality: monoclonal
Clone: EPR7494
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR