Anti-Cystatin A Rabbit Monoclonal Antibody [clone: EPR6941]

Supplier: Abcam
ABCAAB128948-100EA 545 EUR
ABCAAB128948-100
Anti-Cystatin A Rabbit Monoclonal Antibody [clone: EPR6941]
Antibodies
Type: Primary
Antigen: Cystatin A
Clonality: monoclonal
Clone: EPR6941
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR