Anti-RAB3D Rabbit Monoclonal Antibody [clone: EPR8106]
Supplier: Abcam
ABCAAB128997-100EA 515 EUR
ABCAAB128997-100
Anti-RAB3D Rabbit Monoclonal Antibody [clone: EPR8106]
Antibodies
Type: Primary
Antigen: RAB3D (RAB3D, member RAS oncogene family)
Clonality: monoclonal
Clone: EPR8106
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now