Anti-ALDH5A1 Rabbit Monoclonal Antibody [clone: EPR7794]

Supplier: Abcam
ABCAAB129017-100EA 525 EUR
ABCAAB129017-100
Anti-ALDH5A1 Rabbit Monoclonal Antibody [clone: EPR7794]
Antibodies
Type: Primary
Antigen: ALDH5A1 (aldehyde dehydrogenase 5 family, member A1)
Clonality: monoclonal
Clone: EPR7794
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR