Anti-DPYSL2 Rabbit Monoclonal Antibody [clone: EPR7792]

Supplier: Abcam
ABCAAB129082-100EA 525 EUR
ABCAAB129082-100
Anti-DPYSL2 Rabbit Monoclonal Antibody [clone: EPR7792]
Antibodies
Type: Primary
Antigen: DPYSL2 (dihydropyrimidinase-like 2)
Clonality: monoclonal
Clone: EPR7792
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR