Anti-IMPDH2 Rabbit Monoclonal Antibody [clone: EPR8365]

Supplier: Abcam
ABCAAB129165-100EA 535 EUR
ABCAAB129165-100
Anti-IMPDH2 Rabbit Monoclonal Antibody [clone: EPR8365]
Antibodies
Type: Primary
Antigen: IMPDH2 (IMP (inosine 5'-monophosphate) dehydrogenase 2)
Clonality: monoclonal
Clone: EPR8365
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR