Anti-DIAPH1 Rabbit monoclonal antibody unconjugated [clone: EPR7948]

Supplier: Abcam
ABCAAB129167-100EA 530 EUR
ABCAAB129167-100
Anti-DIAPH1 Rabbit monoclonal antibody unconjugated [clone: EPR7948]
Antibodies
Type: Primary
Antigen: DIAPH1 (diaphanous-related formin 1)
Clonality: monoclonal
Clone: EPR7948
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR