Anti-NCL Rabbit Monoclonal Antibody [clone: EPR7952]

Supplier: Abcam
ABCAAB129200-100EA 525 EUR
ABCAAB129200-100
Anti-NCL Rabbit Monoclonal Antibody [clone: EPR7952]
Antibodies
Type: Primary
Antigen: NCL (nucleolin)
Clonality: monoclonal
Clone: EPR7952
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR