Anti-PRDX3 Rabbit Monoclonal Antibody [clone: EPR8114]

Supplier: Abcam
ABCAAB129206-100EA 585 EUR
ABCAAB129206-100
Anti-PRDX3 Rabbit Monoclonal Antibody [clone: EPR8114]
Antibodies
Type: Primary
Antigen: PRDX3 (peroxiredoxin 3)
Clonality: monoclonal
Clone: EPR8114
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR