Anti-TRAF1 Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB129279-50EA 560 EUR
ABCAAB129279-50
Anti-TRAF1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TRAF1 (TNF receptor-associated factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR