Anti-CHST9 Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB129779-100EA 510 EUR
ABCAAB129779-100
Anti-CHST9 Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: CHST9 (carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR