Anti-WWOX Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB129881-100EA 550 EUR
ABCAAB129881-100
Anti-WWOX Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: WWOX (WW domain containing oxidoreductase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR