Anti-Dopamine Transporter Goat polyclonal antibody

Supplier: Abcam
ABCAAB130140-200EA 550 EUR
ABCAAB130140-200
Anti-Dopamine Transporter Goat polyclonal antibody
Type: Primary
Antigen: Dopamine Transporter
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR