Anti-XRCC5 Rabbit Monoclonal Antibody [clone: SP130]

Supplier: Abcam
ABCAAB130752-100EA 570 EUR
ABCAAB130752-100
Anti-XRCC5 Rabbit Monoclonal Antibody [clone: SP130]
Antibodies
Type: Primary
Antigen: XRCC5 (X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining))
Clonality: monoclonal
Clone: SP130
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR