Anti-DAPP1 Rabbit monoclonal antibody unconjugated [clone: EPR4993]

Supplier: Abcam
ABCAAB131212-100EA 495 EUR
ABCAAB131212-100
Anti-DAPP1 Rabbit monoclonal antibody unconjugated [clone: EPR4993]
Antibodies
Type: Primary
Antigen: DAPP1 (dual adaptor of phosphotyrosine and 3-phosphoinositides)
Clonality: monoclonal
Clone: EPR4993
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR