Anti-RNF8 Rabbit Monoclonal Antibody [clone: EPR6312]

Supplier: Abcam
ABCAAB131221-100EA 550 EUR
ABCAAB131221-100
Anti-RNF8 Rabbit Monoclonal Antibody [clone: EPR6312]
Antibodies
Type: Primary
Antigen: RNF8 (ring finger protein 8, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone: EPR6312
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR