Anti-TRIM32 Rabbit Monoclonal Antibody [clone: EP6355]

Supplier: Abcam
ABCAAB131223-100EA 560 EUR
ABCAAB131223-100
Anti-TRIM32 Rabbit Monoclonal Antibody [clone: EP6355]
Antibodies
Type: Primary
Antigen: TRIM32 (tripartite motif containing 32)
Clonality: monoclonal
Clone: EP6355
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR