Anti-CD84 Rabbit Monoclonal Antibody [clone: EPR8325]

Supplier: Abcam
ABCAAB131256-100EA 575 EUR
ABCAAB131256-100
Anti-CD84 Rabbit Monoclonal Antibody [clone: EPR8325]
Antibodies
Type: Primary
Antigen: CD84 (CD84 molecule)
Clonality: monoclonal
Clone: EPR8325
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR