Anti-SMPD2 Rabbit Monoclonal Antibody [clone: EPR6718]

Supplier: Abcam
ABCAAB131330-100EA 520 EUR
ABCAAB131330-100
Anti-SMPD2 Rabbit Monoclonal Antibody [clone: EPR6718]
Antibodies
Type: Primary
Antigen: SMPD2 (sphingomyelin phosphodiesterase 2, neutral membrane (neutral sphingomyelinase))
Clonality: monoclonal
Clone: EPR6718
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR