Anti-CD34 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 581]

Supplier: Abcam
ABCAAB131589-50EA 550 EUR
ABCAAB131589-50
Anti-CD34 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 581]
Antibodies
Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: monoclonal
Clone: 581
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR