Anti-ZIC3 Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB133153-200EA 510 EUR
ABCAAB133153-200
Anti-ZIC3 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZIC3 (Zic family member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR