Anti-XPO4 Rabbit Monoclonal Antibody [clone: EPR4442]

Supplier: Abcam
ABCAAB133237-100EA 495 EUR
ABCAAB133237-100
Anti-XPO4 Rabbit Monoclonal Antibody [clone: EPR4442]
Antibodies
Type: Primary
Antigen: XPO4 (exportin 4)
Clonality: monoclonal
Clone: EPR4442
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR