Anti-CYB5R3 Rabbit Monoclonal Antibody [clone: EPR3868]

Supplier: Abcam
ABCAAB133247-100EA 475 EUR
ABCAAB133247-100
Anti-CYB5R3 Rabbit Monoclonal Antibody [clone: EPR3868]
Antibodies
Type: Primary
Antigen: CYB5R3 (cytochrome b5 reductase 3)
Clonality: monoclonal
Clone: EPR3868
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR