Anti-MAPKAPK2 Rabbit Monoclonal Antibody [clone: EPR5357]

Supplier: Abcam
ABCAAB133279-100EA 580 EUR
ABCAAB133279-100
Anti-MAPKAPK2 Rabbit Monoclonal Antibody [clone: EPR5357]
Antibodies
Type: Primary
Antigen: MAPKAPK2 (mitogen-activated protein kinase-activated protein kinase 2)
Clonality: monoclonal
Clone: EPR5357
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR