Anti-ALDH2 Rabbit Monoclonal Antibody [clone: EPR4494]

Supplier: Abcam
ABCAAB133306-100EA 530 EUR
ABCAAB133306-100
Anti-ALDH2 Rabbit Monoclonal Antibody [clone: EPR4494]
Antibodies
Type: Primary
Antigen: ALDH2 (aldehyde dehydrogenase 2 family (mitochondrial))
Clonality: monoclonal
Clone: EPR4494
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR