Anti-CD4 Rabbit Monoclonal Antibody [clone: EPR3942]

Supplier: Abcam
ABCAAB133313-100EA 545 EUR
ABCAAB133313-100
Anti-CD4 Rabbit Monoclonal Antibody [clone: EPR3942]
Antibodies
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: monoclonal
Clone: EPR3942
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR