Anti-FGG Rabbit Monoclonal Antibody [clone: EPR3085]

Supplier: Abcam
ABCAAB133339-100EA 545 EUR
ABCAAB133339-100
Anti-FGG Rabbit Monoclonal Antibody [clone: EPR3085]
Antibodies
Type: Primary
Antigen: FGG (fibrinogen gamma chain)
Clonality: monoclonal
Clone: EPR3085
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR