Anti-NCAM1 Rabbit Monoclonal Antibody [clone: EPR2566]

Supplier: Abcam
ABCAAB133345-100EA 520 EUR
ABCAAB133345-100
Anti-NCAM1 Rabbit Monoclonal Antibody [clone: EPR2566]
Antibodies
Type: Primary
Antigen: NCAM1 (neural cell adhesion molecule 1)
Clonality: monoclonal
Clone: EPR2566
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR