Imprimer

Anti-UBE1L Rabbit Monoclonal Antibody

Fournisseur: Abcam
ABCAAB133479-100EA 510 EUR
ABCAAB133479-100
Anti-UBE1L Rabbit Monoclonal Antibody
Anticorps
Type: Primary
Antigen: UBE1L
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR