Anti-HMGB2 Rabbit Monoclonal Antibody

Supplier: Abcam
ABCAAB133540-100EA 490 EUR
ABCAAB133540-100
Anti-HMGB2 Rabbit Monoclonal Antibody
Antibodies
Type: Primary
Antigen: HMGB2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR