Anti-DPP3 Rabbit Monoclonal Antibody [clone: EPR9021]

Supplier: Abcam
ABCAAB133735-100EA 495 EUR
ABCAAB133735-100
Anti-DPP3 Rabbit Monoclonal Antibody [clone: EPR9021]
Antibodies
Type: Primary
Antigen: DPP3 (dipeptidyl-peptidase 3)
Clonality: monoclonal
Clone: EPR9021
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR