Anti-PARP4 Rabbit Monoclonal Antibody [clone: EPR8230]

Supplier: Abcam
ABCAAB133745-100EA 500 EUR
ABCAAB133745-100
Anti-PARP4 Rabbit Monoclonal Antibody [clone: EPR8230]
Antibodies
Type: Primary
Antigen: PARP4 (poly (ADP-ribose) polymerase family, member 4)
Clonality: monoclonal
Clone: EPR8230
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR