Anti-CFB Rabbit monoclonal antibody unconjugated [clone: EPR9288]

Supplier: Abcam
ABCAAB133765-100EA 525 EUR
ABCAAB133765-100
Anti-CFB Rabbit monoclonal antibody unconjugated [clone: EPR9288]
Antibodies
Type: Primary
Antigen: CFB (complement factor B)
Clonality: monoclonal
Clone: EPR9288
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR