Anti-IgG2 Rabbit Monoclonal Antibody [clone: EPR4418]

Supplier: Abcam
ABCAAB134050-100EA 580 EUR
ABCAAB134050-100
Anti-IgG2 Rabbit Monoclonal Antibody [clone: EPR4418]
Antibodies
Type: Primary
Antigen: IgG2
Clonality: monoclonal
Clone: EPR4418
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR