Anti-PDGF Receptor alpha Rabbit Monoclonal Antibody [clone: EP2478]

Supplier: Abcam
ABCAAB134068-100EA 500 EUR
ABCAAB134068-100
Anti-PDGF Receptor alpha Rabbit Monoclonal Antibody [clone: EP2478]
Antibodies
Type: Primary
Antigen: PDGF Receptor alpha
Clonality: monoclonal
Clone: EP2478
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR