Anti-UBR5 Rabbit Monoclonal Antibody [clone: EPR4841]

Supplier: Abcam
ABCAAB134089-100EA 505 EUR
ABCAAB134089-100
Anti-UBR5 Rabbit Monoclonal Antibody [clone: EPR4841]
Antibodies
Type: Primary
Antigen: UBR5 (ubiquitin protein ligase E3 component n-recognin 5)
Clonality: monoclonal
Clone: EPR4841
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR