Anti-MADD Rabbit Monoclonal Antibody [clone: EPR4919]

Supplier: Abcam
ABCAAB134117-100EA 505 EUR
ABCAAB134117-100
Anti-MADD Rabbit Monoclonal Antibody [clone: EPR4919]
Antibodies
Type: Primary
Antigen: MADD (MAP-kinase activating death domain)
Clonality: monoclonal
Clone: EPR4919
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR