Anti-SLC17A8 Mouse monoclonal antibody [clone: S34-34]

Supplier: Abcam
ABCAAB134310-100EA 495 EUR
ABCAAB134310-100
Anti-SLC17A8 Mouse monoclonal antibody [clone: S34-34]
Type: Primary
Antigen: SLC17A8 (solute carrier family 17 (vesicular glutamate transporter), member 8)
Clonality: monoclonal
Clone: S34-34
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR