Anti-NCAM1 Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: MEM-188]

Supplier: Abcam
ABCAAB134333-50EA 550 EUR
ABCAAB134333-50
Anti-NCAM1 Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: MEM-188]
Antibodies
Type: Primary
Antigen: NCAM1 (neural cell adhesion molecule 1)
Clonality: monoclonal
Clone: MEM-188
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR