Anti-CD28 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: CD28.2]

Supplier: Abcam
ABCAAB134418-50EA 550 EUR
ABCAAB134418-50
Anti-CD28 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: CD28.2]
Antibodies
Type: Primary
Antigen: CD28 (CD28 molecule)
Clonality: monoclonal
Clone: CD28.2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR