Anti-PI4 kinase beta Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB134756-100EA 560 EUR
ABCAAB134756-100
Anti-PI4 kinase beta Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PI4 kinase beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR